Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Positive Control - WB: human HCCP tissue, rat liver tissue, mouse liver tissue. IHC: rat brain tissue, human lung cancer tissue. ICC/IF: U20S cell, U20S cell. Flow: A431 cell.
The protein encoded by this gene is a member of subfamily 7 in the aldehyde dehydrogenase gene family. These enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. This particular member has homology to a previously described protein from the green garden pea, the 26g pea turgor protein. It is also involved in lysine catabolism that is known to occur in the mitochondrial matrix. Recent reports show that this protein is found both in the cytosol and the mitochondria, and the two forms likely arise from the use of alternative translation initiation sites. An additional variant encoding a different isoform has also been found for this gene. Mutations in this gene are associated with pyridoxine-dependent epilepsy. Several related pseudogenes have also been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 26g turgor protein homolog; aldehyde dehydrogenase 7 family, member A1; Aldehyde dehydrogenase family 7 member A1; aldehyde dehydrogenase family 7, member A1; Alpha-AASA dehydrogenase; Alpha-aminoadipic semialdehyde dehydrogenase; Antiquitin 1; Antiquitin-1; Antiquitin1; Betaine aldehyde dehydrogenase; Delta1-piperideine-6-carboxylate dehydrogenase; delta1-piperideine-6-carboxylate dehydrogenease; P6c dehydrogenase
Gene Aliases: Ald7a1; ALDH7A1; antiquitin; ATQ1; D18Wsu181e; EPD; PDE
UniProt ID: (Human) P49419, (Rat) Q64057, (Mouse) Q9DBF1
Entrez Gene ID: (Human) 501, (Rat) 291450, (Mouse) 110695
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support