Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The antibody detects endogenous levels of total ALDH8A1 protein.
This protein belongs to the aldehyde dehydrogenases family of proteins. It plays a role in a pathway of 9-cis-retinoic acid biosynthesis in vivo. This enzyme converts 9-cis-retinal into the retinoid X receptor ligand 9-cis-retinoic acid, and has approximately 40-fold higher activity with 9-cis-retinal than with all-trans-retinal. Therefore, it is the first known aldehyde dehydrogenase to show a preference for 9-cis-retinal relative to all-trans-retinal. Two transcript variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-aminomuconic semialdehyde dehydrogenase; Aldehyde dehydrogenase 12; aldehyde dehydrogenase 8 family, member A1; Aldehyde dehydrogenase family 8 member A1; aldehyde dehydrogenase family protein; Retinal dehydrogenase 4; retinal dehydrogenase type 4
Gene Aliases: ALDH12; ALDH8A1; DJ352A20.2; Raldh4
UniProt ID: (Human) Q9H2A2, (Mouse) Q8BH00
Entrez Gene ID: (Human) 64577, (Mouse) 237320
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support