Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HNYGYSIMG LVHGPNHPLV LLAKWYEKFF GRLSHLNLCV TNAMREDLAD NWHIRAVTVY DKPASFFKET PLDLQHRLFM KLGSMHSPFR ARSEPEDPVT ERSAFTERDA GSGLVTRLRE RPALLVSSTS WTEDEDFSIL LAALEKFEQL TLDGHNLPSL VCVITGKGPM REYYSRLIHQ KHFQHIQVCT PWLEAEDYPL LLGSADLGVC LHTSSSGLDL PMKVVDMFGC CLPVCAVNFK CLHELVKHEE NGLVFEDSEE LAAQLQMLFS NFPDPAGKLN QFRKNLRESQ QLRWDESWVR TVLPLVMDT (157-464 aa encoded by BC031095)
The enzyme encoded by this gene catalyzes the first mannosylation step in the biosynthesis of lipid-linked oligosaccharides. This gene is mutated in congenital disorder of glycosylation type Ik.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Argonaute; Argonaute-like; asparagine-linked glycosylation 1 homolog (S. cerevisiae, beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1 homolog (yeast, beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1, beta-1,4-mannosyltransferase homolog; Asparagine-linked glycosylation protein 1 homolog; beta-1,4 mannosyltransferase; Beta-1,4-mannosyltransferase; Chitobiosyldiphosphodolichol beta-mannosyltransferase; GDP-Man:GlcNAc2-PP-dolichol mannosyltransferase; GDP-mannose-dolichol diphosphochitobiose mannosyltransferase; Mannosyltransferase-1; MT-1
Gene Aliases: ALG1; CDG1K; hMat-1; HMAT1; HMT-1; HMT1; Mat-1; MT-1; PSEC0061; UNQ861/PRO1870
UniProt ID: (Human) Q9BT22, (Mouse) Q921Q3
Entrez Gene ID: (Human) 56052, (Rat) 360475, (Mouse) 208211
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support