Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Isoform 1: Possible multifunctional enzyme with both glycosyltransferase and deubiquitinase activities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Asparagine-linked glycosylation 13 homolog; Glycosyltransferase 28 domain-containing protein 1; hematopoietic stem/progenitor cells protein MDS031; N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase; Putative bifunctional UDP-N-acetylglucosamine transferase and deubiquitinase ALG13; tudor domain containing 13; UDP-N-acetylglucosamine transferase subunit ALG13 homolog
Gene Aliases: ALG13; CDG1S; CXorf45; EIEE36; GLT28D1; MDS031; TDRD13; YGL047W
UniProt ID: (Human) Q9NP73
Entrez Gene ID: (Human) 79868
Molecular Function:
cysteine protease
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support