Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKCVFVTVGT TSFDDLIACV SAPDSLQKIE SLGYNRLILQ IGRGTVVPEP FSTESFTLDV YRYKDSLKED IQKADLVISH AGAGSCLETL EKGKPLVVVI NEKLMNNHQL ELAKQLHKEG HLFYCTCSTL PGLLQSMDLS TLKCYPPGQP EKFSAFLDKV VGLQK
Isoform 1: Possible multifunctional enzyme with both glycosyltransferase and deubiquitinase activities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Asparagine-linked glycosylation 13 homolog; Glycosyltransferase 28 domain-containing protein 1; hematopoietic stem/progenitor cells protein MDS031; N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase; tudor domain containing 13; UDP-N-acetylglucosamine transferase subunit ALG13; UDP-N-acetylglucosamine transferase subunit ALG13 homolog
Gene Aliases: ALG13; CDG1S; CXorf45; EIEE36; GLT28D1; MDS031; TDRD13; YGL047W
UniProt ID: (Human) Q9NP73
Entrez Gene ID: (Human) 79868
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support