Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
This gene encodes a member of the ALG3 family. The encoded protein catalyzes the addition of the first dol-P-Man derived mannose in an alpha 1,3 linkage to Man5GlcNAc2-PP-Dol. Defects in this gene have been associated with congenital disorder of glycosylation type Id (CDG-Id) characterized by abnormal N-glycosylation. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: asparagine-linked glycosylation 3 homolog (S. cerevisiae, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 3 homolog (yeast, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 3, alpha-1,3- mannosyltransferase homolog; Asparagine-linked glycosylation protein 3 homolog; carbohydrate deficient glycoprotein syndrome type IV; dol-P-Man dependent alpha(1-3)-mannosyltransferase; dol-P-Man dependent alpha-1,3- mannosyltransferase; Dol-P-Man-dependent alpha(1-3)-mannosyltransferase; Dol-P-Man:Man(5)GlcNAc(2)-PP-Dol alpha-1,3-mannosyltransferase; dol-P-Man:Man(5)GlcNAc(2)-PP-Dol3-mannosyltransferase; Dolichyl-P-Man:Man(5)GlcNAc(2)-PP-dolichyl mannosyltransferase; Dolichyl-phosphate-mannose--glycolipid alpha-mannosyltransferase; Not56-like protein
Gene Aliases: ALG3; CDG1D; CDGS4; CDGS6; D16Ertd36e; NOT; Not56; NOT56L
UniProt ID: (Human) Q92685
Entrez Gene ID: (Human) 10195
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support