Search Thermo Fisher Scientific
Immunogen sequence: MEPNPHWHP VLRTLKNRIE ENTGHTFNSL LCNLYRNEKD SVDWHSDDEP SLGRCPIIAS LSFGATRTFE MRKKPPPEEN GDYTYVERVK IPLDHGTLLI MEGATQADWQ HRVPKEYHSR EPRVNLTFRT VYPDPRGAPW (148-286 aa encoded by BC015155)
Dioxygenase that repairs alkylated DNA containing 1- methyladenine (1meA) and 3-methylcytosine (3meC) by oxidative demethylation. Has a strong preference for single-stranded DNA. Able to process alkylated 3mC within double-stranded regions via its interaction with ASCC3, which promotes DNA unwinding to generate single-stranded substrate needed for ALKHB3. May also act on RNA. Requires molecular oxygen, alpha-ketoglutarate and iron.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AlkB homolog 3; alkB homolog 3, alpha-ketoglutarate-dependent dioxygenase; alkB, alkylation repair homolog 3; Alkylated DNA repair protein alkB homolog 3; Alpha-ketoglutarate-dependent dioxygenase alkB homolog 3; DEPC 1; DEPC-1; FLJ43614; hABH3; m MGC118790; mAbh3; MGC118792; MGC118793; MGC134125; Prostate cancer antigen 1; prostate cancer antigen-1; RP23-375N21.4
Gene Aliases: 1700108H04Rik; 1810020C19Rik; ABH3; ALKBH3; DEPC-1; DEPC1; hABH3; mABH3; PCA1
UniProt ID: (Human) A6NDJ1, (Rat) Q5XIC8, (Mouse) Q8K1E6
Entrez Gene ID: (Human) 221120, (Rat) 362169, (Mouse) 69113
Molecular Function: oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support