Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Oxygenase and 14,15-leukotriene A4 synthase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 12(S)-lipoxygenase; 12LOX; 12S LOX; 12S-lipoxygenase; 12S-LOX; Arachidonate (12S)-lipoxygenase; Arachidonate (15S)-lipoxygenase; arachidonate 12-lipoxygenase; Linoleate (13S)-lipoxygenase; Lipoxin synthase 12-LO; platelet 12-LOX; platelet-type 12-lipoxygenase; Platelet-type lipoxygenase 12; Polyunsaturated fatty acid lipoxygenase ALOX12
Gene Aliases: 12-LOX; 12LO; 12S-LOX; ALOX12; LOG12
UniProt ID: (Human) P18054
Entrez Gene ID: (Human) 239
Molecular Function: oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support