Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
ALOX15 converts arachidonic acid to 15S-hydroperoxyeicosatetraenoic acid. The protein also acts on C-12 of arachidonate as well as on linoleic acid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 12-LOX; 12/15-lipoxygenase; 12/15-LOX; 15-lipooxygenase-1; 15-LOX; 15-LOX2; Arachidonate 12-lipoxygenase, leukocyte-type; Arachidonate 15-lipoxygenase; Arachidonate omega-6 lipoxygenase; Hepoxilin A3 synthase Alox15; Linoleate 13S-lipoxygenase; Polyunsaturated fatty acid lipoxygenase ALOX15
Gene Aliases: 12-LOX; 15-LOX; 15-LOX-1; 15LOX-1; Alox12; Alox12l; ALOX15; LOG15
UniProt ID: (Human) P16050, (Rat) Q02759
Entrez Gene ID: (Human) 246, (Rat) 81639
Molecular Function: oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support