Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
This gene encodes a protein which, with Arachidonate 5-lipoxygenase-activating protein, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the plasma membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the cell membrane and inhibit 5-lipoxygenase activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5 - Lipoxygenase; 5-lipoxygenase; 5-LO; Arachidonate 5-lipoxygenase; arachidonic 5-lipoxygenase alpha-10 isoform; arachidonic 5-lipoxygenase delta-10-13 isoform; arachidonic 5-lipoxygenase delta-13 isoform; arachidonic 5-lipoxygenase delta-p10 isoform; arachidonic acid 5-lipoxygenase; leukotriene A4 synthase; LOX-5; lox5; Polyunsaturated fatty acid 5-lipoxygenase; RP11-67C2.3
Gene Aliases: 5-LO; 5-LOX; 5LO; 5LPG; 5LX; AI850497; ALOX5; F730011J02; LOG5; LOX5A
UniProt ID: (Human) P09917, (Rat) P12527, (Mouse) P48999
Entrez Gene ID: (Human) 240, (Rat) 25290, (Mouse) 11689
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support