Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDSKKRSSTE AEGSKERGLV HIWQAGSFPI TPERLPGWGG KTVLQAALGV KHGVLLTEDG EVYSFGTLPW RSGPVEICPS SPILENALVG QYVITVATGS FHSGAVTDNG VAYMWGENSA GQCAVANQQY VPEPNPVSIA DSEASPLLAV RILQLACGEE HTLALSISRE IWAWGTGCQL GLITTAFPVT KPQKVEHLAG RVVLQVACGA FHSLALVQCL PSQDLKPVPE RCNQCSQLLI TMTDKEDHVI ISDSHCCPLG VTLTESQAEN HASTALSPST ETLDRQEEVF ENTLVANDQS VATELNAVSA QITSSDAMSS QQNVMGTTEI SSARNIPSYP DTQAVNEYLR KLSDHSVRED SEHGEKPVPS QPLLEEAIPN LHSPPTTSTS ALNSLVVSCA SAVGVRVAAT YEAGALSLKK VMNFYSTTPC ETGAQAGSSA IGPEGLKDSR EEQVKQESMQ GKKSSSLVDI REEETEGGSR RLSLPGLLSQ VSPRLLRKAA RVKTRTVVLT PTYSGEADAL LPSLRIEVWT WGKGKEGQLG HGDVLPRLQP LCVKCLDGKE VIHLEAGGYH SLALTAKSQV YSWGSNTFGQ LGHSDFPTTV PRLAKISSEN GVWSIAAGRD YSLFLVDTED FQPGLYYSGR QDPTEGDNLP ENHSGSKTPV LLSCSKLGYI SRVTAGKDSY LALVDKNIMG YIASLHELAT TERRFYSKLS DIKSQILRPL LSLENLGTTT TVQLLQEVAS RFSKLCYLIG QHGASLSSFL HGVKEARSLV ILKHSSLFLD SYTEYCTSIT NFLVMGGFQL LAKPAIDFLN KNQELLQDLS EVNDENTQLM EILNTLFFLP IRRLHNYAKV LLKLATCFEV ASPEYQKLQD SSSCYECLAL HLGRKRKEAE YTLGFWKTFP GKMTDSLRKP ERRLLCESSN RALSLQHAGR FSVNWFILFN DALVHAQFST HHVFPLATLW AEPLSEEAGG VNGLKITTPE EQFTLISSTP QEKTKWLRAI SQAVDQALRG MSDLPPYGSG SSVQRQEPPI SRSAKYTFYK DPRLKDATYD GRWLSGKPHG RGVLKWPDGK MYSGMFRNGL EDGYGEYRIP NKAMNKEDHY VGHWKEGKMC GQGVYSYASG EVFEGCFQDN MRHGHGLLRS GKLTSSSPSM FIGQWVMDKK AGYGVFDDIT RGEKYMGMWQ DDVCQGNGVV VTQFGLYYEG NFHLNKMMGN GVLLSEDDTI YEGEFSDDWT LSGKVGN
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alsin; amyotrophic lateral sclerosis 2 (juvenile); Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 6 protein; Amyotrophic lateral sclerosis 2 protein; FLJ31851; MGC87187
Gene Aliases: ALS2; ALS2CR6; ALSJ; IAHSP; KIAA1563; PLSJ
UniProt ID: (Human) Q96Q42
Entrez Gene ID: (Human) 57679
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support