Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: PSQDLKPVPE RCNQCSQLLI TMTDKEDHVI ISDSHCCPLG VTLTESQAEN HASTALSPST ETLDRQEEVF ENTLVANDQS VATELNAVSA QITSSDAMSS
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alsin; amyotrophic lateral sclerosis 2 (juvenile); Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 6 protein; Amyotrophic lateral sclerosis 2 protein; FLJ31851; MGC87187
Gene Aliases: ALS2; ALS2CR6; ALSJ; IAHSP; KIAA1563; PLSJ
UniProt ID: (Human) Q96Q42
Entrez Gene ID: (Human) 57679
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support