Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Western blot applications, this antibody detects a band at ~34kDa. A suggested positive control is human heart tissue lysate.
PA5-20036 can be used with blocking peptide PEP-0155.
This gene encodes a protein that belongs to the serine/threonine protein kinase STE20 subfamily. One of the active site residues in the protein kinase domain of this protein is altered, and it is thus a pseudokinase. This protein is a component of a complex involved in the activation of serine/threonine kinase 11, a master kinase that regulates cell polarity and energy-generating metabolism. This complex regulates the relocation of this kinase from the nucleus to the cytoplasm, and it is essential for G1 cell cycle arrest mediated by this kinase. The protein encoded by this gene can also interact with the X chromosome-linked inhibitor of apoptosis protein, and this interaction enhances the anti-apoptotic activity of this protein via the JNK1 signal transduction pathway. Two pseudogenes, located on chromosomes 1 and 7, have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 2; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 2 protein; CALS-21; ILP-interacting protein; ILP-interacting protein ILPIPA; MGC102916; Pseudokinase ALS2CR2; STE20-related kinase adapter protein beta; STRAD beta
Gene Aliases: ALS2CR2; CALS-21; ILPIP; ILPIPA; PAPK; PRO1038; STRADB
UniProt ID: (Human) Q9C0K7
Entrez Gene ID: (Human) 55437
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support