Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
AMDHD1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amidohydrolase domain-containing protein 1; Probable imidazolonepropionase
Gene Aliases: AMDHD1; HMFT1272
UniProt ID: (Human) Q96NU7
Entrez Gene ID: (Human) 144193
Molecular Function: metalloprotease protease protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support