Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LYILTHKRI LKFLKLFITE FPKPEFMSKS LEELQIGTYA NIAMVRTTTP VYVALGIFVQ HRVSALPVVD EKGRVVDIYS KFDVINLAAE KTYNNLDVSV TKALQHRSHY FEGVLKCYLH ETLETIINRL VEAEVHRLVV VDENDVVKGI VSLSDILQA (164-321 aa encoded by BC000358)
The protein encoded by this gene is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This subunit is one of the gamma regulatory subunits of AMPK. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-AMP-activated protein kinase subunit gamma-1; 5'-AMP-activated protein kinase, gamma-1 subunit; ampk gamma; AMPK gamma-1 chain; AMPK gamma1; protein kinase, AMP-activated, gamma 1 non-catalytic subunit
Gene Aliases: AMPKG; PRKAG1
UniProt ID: (Human) P54619
Entrez Gene ID: (Human) 5571
Molecular Function: kinase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support