Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In direct ELISAs, approximately 100% cross-reactivity with recombinant human (rh) ANGPTL2 is observed, and less than 1% cross-reactivity with rhAngiopoietin-1, rhAngiopoietin-2, recombinant mouse (rm) Angiopoietin-3, rhAngiopoietin-4, rmAngiopoietin-6, and rmAngiopoietin-7 is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: angiopoietin related protein 2; Angiopoietin-like protein 2; Angiopoietin-related protein 2; MGC8889
Gene Aliases: AI593246; Angptl2; Arp2; AW260363
UniProt ID: (Mouse) Q9R045
Entrez Gene ID: (Mouse) 26360
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support