Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HLVAITGNVP NAIPENKDLV FSTWDHKAKG HFNCPEGYSG GWWWHDECGE NNLNGKYNKP RAKSKPERRR GLSWKSQNGR LYSIKSTKML IHPTDSESFE
This gene encodes a member of a family of secreted proteins that function in angiogenesis. The encoded protein, which is expressed predominantly in the liver, is further processed into an N-terminal coiled-coil domain-containing chain and a C-terminal fibrinogen chain. The N-terminal chain is important for lipid metabolism, while the C-terminal chain may be involved in angiogenesis. Mutations in this gene cause familial hypobetalipoproteinemia type 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ANG-5; angiopoietin 5; Angiopoietin-5; angiopoietin-like 3; Angiopoietin-like protein 3; Angiopoietin-related protein 3
Gene Aliases: ANG-5; ANGPT5; ANGPTL3; ANL3; FHBL2; UNQ153/PRO179
UniProt ID: (Human) Q9Y5C1
Entrez Gene ID: (Human) 27329
Molecular Function:
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support