Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: GSTSDTSRML DPAPEPQRDQ TQRQQEPMAS PMPAGHPAVP TKPVGPWQDC AEARQAGHEQ SGVYELRVGR HVVSVWCEQQ LEGGGWTVI
May play a role in the wound healing process. May promote epidermal proliferation, remodeling and regeneration. May promote the chemotactic activity of endothelial cells and induce neovascularization. May counteract high-fat diet-induced obesity and related insulin resistance through increased energy expenditure.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: angiopoietin-like 6; Angiopoietin-like protein 6; Angiopoietin-related growth factor; Angiopoietin-related protein 5; Angiopoietin-related protein 6
Gene Aliases: AGF; ANGPTL6; ARP5; UNQ152/PRO178
UniProt ID: (Human) Q8NI99
Entrez Gene ID: (Human) 83854
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support