Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
Application for antibody includes Stable Isotope Standards and Capture by Anti-Peptide Antibodies (SISCAPA).
Ankyrins are a family of proteins that link the integral membrane proteins to the underlying spectrin-actin cytoskeleton and play key roles in activities such as cell motility, activation, proliferation, contact and the maintenance of specialized membrane domains. Multiple isoforms of ankyrin with different affinities for various target proteins are expressed in a tissue-specific, developmentally regulated manner. Most ankyrins are typically composed of three structural domains: an amino-terminal domain containing multiple ankyrin repeats; a central region with a highly conserved spectrin binding domain; and a carboxy-terminal regulatory domain which is the least conserved and subject to variation. Ankyrin 1, the prototype of this family, was first discovered in the erythrocytes, but since has also been found in brain and muscles. Mutations in erythrocytic ankyrin 1 have been associated in approximately half of all patients with hereditary spherocytosis. Complex patterns of alternative splicing in the regulatory domain, giving rise to different isoforms of ankyrin 1 have been described. Truncated muscle-specific isoforms of ankyrin 1 resulting from usage of an alternate promoter have also been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ANK-1; ankyrin 1, erythrocytic; ankyrin 1, erythroid; Ankyrin-1; Ankyrin-R; Erythrocyte ankyrin; normoblastic anemia; rCG_43073
Gene Aliases: ANK; Ank-1; ANK1; nb; pale; SPH1; SPH2
UniProt ID: (Human) P16157, (Mouse) Q02357
Entrez Gene ID: (Human) 286, (Mouse) 11733, (Rat) 306570
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support