Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Acts as a molecular chaperone for G protein-coupled receptors, regulating their biogenesis and exit from the ER.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat domain-containing protein 13C; RP4-677H15.5
Gene Aliases: ankrd13c; fc57c01; wu:fc57c01; wu:fq56c05; zgc:56077
UniProt ID: (Zebrafish) Q7ZUV0
Entrez Gene ID: (Zebrafish) 325167
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support