Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MGSLNSQLIS TLSSINSVMK SASTEELRNP DFQKTSLGKI TGNYLGYTCK CGGLQSGSPL SSQTSQPEQE VGTSEGKPIS SLDAFPTQEG TLSPVNLTDD
Ankyrins are membrane adaptor molecules that play important roles in the control of cytoskeleton formation by regulating actin polymerization. KANK1 (KN motif and ankyrin repeat domain-containing protein 1), also known as ANKRD15, is a 1,352 amino acid protein that contains at least 12 exons and 5 ANK repeats. It binds to beta-catenin and regulates its subcellular distribution. KANK1 is ubiquitously expressed and localizes to cytoplasm. It may function as a tumor suppressor for renal cell carcinoma. Mutations in this gene cause cerebral palsy spastic quadriplegic type 2, a central nervous system development disorder.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat domain-containing protein 15; Kidney ankyrin repeat-containing protein; KN motif and ankyrin repeat domain-containing protein 1
Gene Aliases: ANKRD15; CPSQ2; KANK; KANK1; KIAA0172
UniProt ID: (Human) Q14678
Entrez Gene ID: (Human) 23189
Molecular Function:
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support