Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: VASEEKQERL QRSENKQPQD SQSYGKKKDA MYGNFMLKKD IAMLKEELYA
Sequence homology: Human: 100%
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat domain-containing protein 18A
Gene Aliases: ANKRD18A; KIAA2015
UniProt ID: (Human) Q8IVF6
Entrez Gene ID: (Human) 253650
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support