Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: VTSQDGSSP LHVAALHGRA DLIPLLLKHG ANAGARNADQ AVPLHLACQQ GHFQVVKCLL DSNAKPNKKD LSGNTPLIYA CSGGHHELVA LLLQHGASIN ASNNKGNTAL HEAVIEKHVF VVELLLLHGA SVQVLNKRQR TAVDCAEQNS KIMELLQVVP SCVASLDDVA ETDRKEYVTV KIRKKWNSKL YDLPDEPFTR QFYFVHSAGQ FKGKTSREIM ARDRSVPNLT EGSLHEPGRQ SVTLRQNNLP AQSGSHAAEK GNSDWPERPG LTQTGPGHRR MLRRHTVEDA VVSQGPEAAG PLSTPQEVSA SRS (739-1050 aa encoded by B C050529)
May be a guanine exchange factor (GEF) for Rab21, Rab32 and Rab38 and regulate endosome dynamics (PubMed:16525121, PubMed:18477474). May regulate the participation of VAMP7 in membrane fusion events; in vitro inhibits VAMP7-mediated SNARE complex formation by trapping VAMP7 in a closed, fusogenically inactive conformation (PubMed:23104059). Involved in peripheral melanosomal distribution of TYRP1 in melanocytes; the function, which probably is implicating vesicle-trafficking, includes cooperation with Rab32, Rab38 and VAMP7. Involved in the regulation of neurite growth; the function seems to require its GEF activity, probably towards Rab21, and VAMP7 but not Rab32/38. Proposed to be involved in Golgi sorting of VAMP7 and transport of VAMP7 vesicles to the cell surface; the function seems to implicate kinesin heavy chain isoform 5 proteins, GOLGA4, RAB21 and MACF1 (PubMed:22705394). Required for the colocalization of VAMP7 and Rab21, probably on TGN sites (PubMed:19745841). Involved in GLUT1 endosome-to-plasma membrane trafficking; the function is dependent of association with VPS29 (PubMed:24856514). Regulates the proper trafficking of melanogenic enzymes TYR, TYRP1 and DCT/TYRP2 to melanosomes in melanocytes. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ankyrin repeat domain 27 (VPS9 domain); Ankyrin repeat domain-containing protein 27; Vps9 domain and ankyrin-repeat-containing protein; VPS9 domain-containing protein; VPS9-ankyrin-repeat protein
Gene Aliases: ANKRD27; PP12899; VARP
UniProt ID: (Human) Q96NW4
Entrez Gene ID: (Human) 84079
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support