Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Plays a role in the DNA damage response (DDR) pathway by regulating postreplication repair of UV-damaged DNA and genomic stability maintenance (PubMed:25931565). The SLF1-SLF2 complex acts to link RAD18 with the SMC5-SMC6 complex at replication-coupled interstrand cross-links (ICL) and DNA double-strand breaks (DSBs) sites on chromatin during DNA repair in response to stalled replication forks (PubMed:25931565). Promotes the recruitment of SLF2 and the SMC5-SMC6 complex to DNA lesions (PubMed:25931565).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ankyrin repeat domain 32; Ankyrin repeat domain-containing protein 32; BRCT domain containing 1; BRCT domain-containing protein 1; Protein BRCTx; SMC5-SMC6 complex localization factor protein 1; Smc5/6 localization factor 1
Gene Aliases: 2700017A04Rik; ANKRD32; AW545819; BRCTD1; BRCTx; C730024G01Rik; SLF1
UniProt ID: (Mouse) Q8R3P9, (Human) Q9BQI6
Entrez Gene ID: (Rat) 294601, (Mouse) 105377, (Human) 84250
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support