Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
Promotions
View available promotion(s)
Promo Code: CI2Z9E .
FIGURE: 1 / 2
Putative regulatory subunit of protein phosphatase 6 (PP6) that may be involved in the recognition of phosphoprotein substrates. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ankyrin repeat domain 33; Ankyrin repeat domain-containing protein 52; CVWG5837; PP6-ARS-C; protein phosphatase 6 ankyrin repeat subunit C; Serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit C; serine/threonine-protein phosphatase 6 regulatory subunit ARS-C
Gene Aliases: 6430544C07; ANKRD33; ANKRD52; G431002C21Rik; RGD1307124
UniProt ID: (Human) Q8NB46, (Mouse) Q8BTI7
Entrez Gene ID: (Human) 283373, (Rat) 362811, (Mouse) 237615
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support