Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YDVNMQDKK YRTPLHLACA NGHTDVVLFL IEQQCKINVR DSENKSPLIK AVQCQNEDCA TILLNFGADP DLRDIRYNTV LHYAVCGQSL SLVEKLLEYE ADLEAKNKDG YTPLLVAVIN NNPKMVKFLL EKGADVNASD NYQRTALILA VSGEPPCLVK LLLQQGVELC YEGIVDSQLR NMFISMVLLH RYPQFTASHG KKKHAK (54-254 aa encoded by BC032799)
ANKRD7 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat domain-containing protein 7; testicular tissue protein Li 19; testis-specific ankyrin motif containing protein; Testis-specific protein TSA806
Gene Aliases: ANKRD7; TSA806
UniProt ID: (Human) Q92527
Entrez Gene ID: (Human) 56311
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support