Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MDMKRRIHLE LRNRTPAAVR ELVLDNCKSN DGKIEGLTAE FVNLEFLSLI NVGLISVSNL PKLPKLKKLE LSENRIFGGL DMLAEKLPNL THLNLSGNKL KDISTLEPLK KLECLKSLDL FNCEVTNLND YRESVFKLLP QLTYLDGYDR EDQEAPDSDA EVDGVDEEEE DEEGEDEEDE DDEDGEEEEF DEEDDEDEDV EGDEDDDEVS EEEEEFGLDE EDEDEDEDEE EEEGGKGEKR KRETDDEGED D
Multifunctional protein working as a cell cycle progression factor as well as a cell survival factor. Required for the progression from the G1 to the S phase. Anti-apoptotic protein which functions as a caspase-3 inhibitor. Has no phosphatase 2A (PP2A) inhibitor activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acidic (leucine-rich) nuclear phosphoprotein 32 family, member B; Acidic leucine-rich nuclear phosphoprotein 32 family member B; Acidic protein rich in leucines; OTTHUMP00000063770; PHAPI2; Putative HLA-DR-associated protein I-2; Silver-stainable protein SSP29; TNF superfamily member 13
Gene Aliases: ANP32B; APRIL; PHAPI2; SSP29
UniProt ID: (Human) Q92688
Entrez Gene ID: (Human) 10541
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support