Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: A431, H1299, HeLa.
Predicted reactivity: Mouse (86%), Rat (84%), Bovine (88%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been discribed and is located approximately 9-kb downstream.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amine oxidase [copper-containing] 3; Amine oxidase copper-containing 3; amine oxidase, copper containing 3 (vascular adhesion protein 1); Copper amine oxidase; HPAO; placenta copper monamine oxidase; Semicarbazide-sensitive amine oxidase; SSAO; VAP-1; Vascular adhesion protein 1
Gene Aliases: AOC3; HPAO; SSAO; VAP-1; VAP1
UniProt ID: (Human) Q16853
Entrez Gene ID: (Human) 8639
Molecular Function:
oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support