Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This gene encodes a subunit of the heterotetrameric adaptor-related protein comlex 1 (AP-1), which belongs to the adaptor complexes medium subunits family. This protein is capable of interacting with tyrosine-based sorting signals. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adaptor protein complex AP-1 mu-2 subunit; Adaptor protein complex AP-1 subunit mu-2; adaptor-related protein complex 1 mu 2 subunit; adaptor-related protein complex 1 mu-2 subunit; Adaptor-related protein complex 1 subunit mu-2; adaptor-related protein complex 1, mu 2 subunit; adaptor-related protein complex AP-1, mu 2 subunit; AP-1 complex subunit mu-2; AP-mu chain family member mu1B; clathrin assembly protein complex 1 medium chain 2; Clathrin assembly protein complex 1 mu-2 medium chain 2; clathrin coat assembly protein AP47 2; clathrin coat associated protein AP47 2; clathrin-associated adaptor medium chain mu2; golgi adaptor AP-1 47 kDa protein; Golgi adaptor HA1/AP1 adaptin mu-2 subunit; HA1 47 kDa subunit 2; Mu-adaptin 2; Mu1B-adaptin
Gene Aliases: [m]1B; AP1-mu2; AP1M2; D9Ertd818e; HSMU1B; MU-1B; MU1B; mu2
UniProt ID: (Human) Q9Y6Q5, (Mouse) Q9WVP1
Entrez Gene ID: (Human) 10053, (Mouse) 11768
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support