Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 9
Immunogen sequence: GSLLKVDVQ GEIRLKSFLP SGSEMRIGLT EEFCVGKSEL RGYGPGIRVD EVSFHSSVNL DEFESHRILR LQPPQGELTV MRYQLSDDLP SPLPFRLFPS VQWDRGSGRL QVYLKLRCDL LSKSQALNVR LHLPLPRGVV SLSQELSSPE QKAELAEGAL RWDLPRVQGG SQLSGLFQMD VPGPPGPPSH GLSTSASPLG LGPASLSFEL PRHTCSGLQV RFLRLAFRPC GNANPHKWVR HLSHSDAYVI RI (203-453 aa encoded by BC018705)
This gene encodes a subunit of the heterotetrameric AP-4 complex. The encoded protein belongs to the adaptor complexes medium subunits family. This AP-4 complex is involved in the recognition and sorting of cargo proteins with tyrosine-based motifs from the trans-Golgi network to the endosomal-lysosomal system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adapter-related protein complex 4 mu-1 subunit; adapter-related protein complex 4 subunit mu-1; adaptor related protein complex 4, mu 1 subunit; Adaptor-related protein complex 4 subunit mu-1; adaptor-related protein complex 4, mu 1 subunit; adaptor-related protein complex AP-4 mu4 subunit; adaptor-related protein complex AP-4, mu 1; AP-4 adapter complex mu subunit; AP-4 adaptor complex mu subunit; AP-4 complex subunit mu-1; ap4m1 {ECO:0000312|RGD:1310233}; Mu subunit of AP-4; Mu-adaptin-related protein 2; mu-adaptin-related protein-2; mu-ARP2; mu4; Mu4-adaptin
Gene Aliases: 4930443L05Rik; AP4M1; Ap4m4; CPSQ3; MU-4; MU-ARP2; MUARP2; SPG50
UniProt ID: (Human) O00189, (Rat) Q2PWT8, (Mouse) Q9JKC7
Entrez Gene ID: (Human) 9179, (Rat) 304344, (Mouse) 11781
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support