Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 3
Positive test controls include: Jurkat, HepG2, HeLa, A-549, Mouse brain, Mouse ovary, Rat brain. The target is usually found in the following locations: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center, spindle.
Immunogen sequence: MLYIGLEYGL TNNSKLAERF FSQALSIAPE DPFVMHEVGV VAFQNGEWKT AEKWFLDALE KIKAIGNEVT VDKWEPLLNN LGHVCRKLKK YAEALDYHRQ ALVLIPQNAS TYSAIGYIHS LMGNFENAVD YFHTALGLRR DDTFSVTMLG HCIEMYIGDS EAYIGADIKD KLKCYDFDVH TMKTLKNIIS PPWDFREFEV EKQTAEETGL TPLETSRKTP DSRPSLEETF EIEMNESDMM LETSMSDHST
The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anaphase-promoting complex subunit 6; anaphase-promoting complex, subunit 6; APC6; CDC16 cell division cycle 16 homolog; CDC16 homolog; CDC16Hs; cell division cycle 16 homolog; Cell division cycle protein 16 homolog; Cyclosome subunit 6; RP11-569D9.4
Gene Aliases: 2700071J12Rik; 2810431D22Rik; ANAPC6; APC6; CDC16; CDC16Hs; CUT9
UniProt ID: (Human) Q7Z651, (Mouse) Q8R349
Entrez Gene ID: (Human) 8881, (Rat) 290875, (Mouse) 69957
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support