Search
Search
FIGURE: 1 / 12
Antibody detects endogenous levels of total CENPS.
APITD1 was identified in the neuroblastoma tumour suppressor candidate region on chromosome 1p36. It contains a TFIID-31 domain, similar to that found in TATA box-binding protein-associated factor, TAF(II)31, which is required for p53-mediated transcription activation. This gene was expressed at very low levels in neuroblastoma tumours, and was shown to reduce cell growth in neuroblastoma cells, suggesting that it may have a role in a cell death pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apoptosis-inducing TAF9-like domain-containing protein 1; Apoptosis-inducing TAF9-like domain-containing protein 1 homolog; CENP-S; Centromere protein S; FANCM-associated histone fold protein 1; FANCM-interacting histone fold protein 1; Fanconi anemia-associated polypeptide of 16 kDa; ortholog of human apoptosis-inducing, TAF9-like domain 1 APITD1
Gene Aliases: 2610040C18Rik; 2810407L01Rik; APITD1; CENP-S; CENPS; FAAP16; MHF1
UniProt ID: (Human) Q8N2Z9, (Mouse) Q9D084
Entrez Gene ID: (Human) 378708, (Mouse) 69928, (Rat) 100534597
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support