Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: QEELLSSQVT QELRALMDET MKELKAYKSE LEEQLTPVAE ETRARLSKEL QAAQARLGAD MEDVCGRLVQ YRGEVQAMLG QSTEELRVRL ASHLRKLRKR LLRDADDLQK RLAVYQAGAR EGAERGLSAI RERLGPLVEQ GRVRAATVGS LAGQPLQERA QAWGERLRAR MEEMGSRTRD RLDEVKEQVA EVRAKLEEQA QQIRLQAEAF QARLKSWFEP LVEDMQRQWA GLVEKVQAAV GTSAAPVPSD NH
Apo E (Apolipoprotein E) plays an important role in the metabolism of lipids in the plasma, and is also is a constituent of various plasma lipoprotein-lipid particles. The protein is synthesized in liver, brain, spleen, and the kidney, and concentrations are high in interstitial fluids, where it plays a role in distributing cholesterol from cells where cholesterol is in excess to cells which require cholesterol. Apo E also plays an important role in the formation of very low density lipoprotein and chylomicrons. The Apo E gene is located on chromosome 19 at position 19q13. 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apo-E; ApoE4; Apolipoprotein; Apolipoprotein E; apolipoprotein E3; apolipoprotein E4; Apoprotein; B2G1; BG; MGC1571
Gene Aliases: AD2; AI255918; APO-E; APOE; LDLCQ5; LPG
UniProt ID: (Human) P02649, (Mouse) P08226
Entrez Gene ID: (Human) 348, (Mouse) 11816
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support