Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MAQLGKLLKE QKYDRQLRLW GDHGQEALES AHVCLINATA TGTEILKNLV LPGIGSFTII DGNQVSGEDA GNNFFLQRSS IGKNRAEAAM EFLQELNSDV SGSFVEESPE NLLDNDPSFF CRFTVVVATQ LPESTSLRLA DVLWNSQIPL LICRTYGLVG YMRIIIKEHP VIESHPDNAL EDLRLDKPFP ELREHFQSYD LDHMEKKDHS HTPWIVIIAK YLAQWYSETN GRIPKTYKEK EDFRDLIRQG ILKNENGAPE DEENFEEAIK NVNTALNTTQ IPSSIEDIFN DDRCINITKQ TPSFWILARA LKEFVAKEGQ GNLPVRGTIP DMIADSGKYI KLQNVYREKA KKDAAAVGNH VAKLLQSIGQ APESISEKEL KLLCSNSAFL RVVRCRSLAE EYGLDTINKD EIISSMDNPD NEIVLYLMLR AVDRFHKQQG RYPGVSNYQV EEDIGKLKSC LTGFLQEYGL SVMVKDDYVH EFCRYGAAEP HTIAAFLGGA AAQEVIKIIT KQFVIFNNTY IYSGMSQTSA TFQL
The protein encoded by this gene binds to the beta-amyloid precursor protein. Beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer's disease. In addition, the encoded protein can form a heterodimer with UBE1C and bind and activate NEDD8, a ubiquitin-like protein. This protein is required for cell cycle progression through the S/M checkpoint. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amyloid beta precursor protein binding protein 1, 59kDa; Amyloid beta precursor protein-binding protein 1, 59 kDa; amyloid beta precursor protein-binding protein 1, 59kD; Amyloid protein-binding protein 1; APP-BP1; NEDD8-activating enzyme E1 regulatory subunit; NEDD8-activating enzyme E1 subunit; Proto-oncogene protein 1; protooncogene protein 1
Gene Aliases: A-116A10.1; APPBP1; HPP1; NAE1; ula-1
UniProt ID: (Human) Q13564
Entrez Gene ID: (Human) 8883
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support