Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The aquaporins are a family of water-selective membrane channels. This gene encodes a member of a subset of aquaporins called the aquaglyceroporins. This protein allows passage of a broad range of noncharged solutes and also stimulates urea transport and osmotic water permeability. This protein may also facilitate the uptake of glycerol in hepatic tissue. The encoded protein may also play a role in specialized leukocyte functions such as immunological response and bactericidal activity. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AQP-9; Aquaglyceroporin-9; Aquaporin-9; neutral solute channel aquaporin 9; Small solute channel 1
Gene Aliases: 1700020I22Rik; AI266899; AQP-9; AQP9; HsT17287; SSC1; T17287
UniProt ID: (Human) O43315, (Mouse) Q9JJJ3, (Rat) P56627
Entrez Gene ID: (Human) 366, (Mouse) 64008, (Rat) 65054
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support