Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KELQQARRD AERQGKKAPG FGGFGSSAVS GGSTAAMITE TIIETDKPKV APAPARPSGP SKALKLGAKG KEVDNFVDKL KSEGETIMSS SMGKRTSEAT KMHAPPINME SVHMKIEEKI TLTCGRDGGL QNMELHGMIM LRISDDKYGR IRLHVENEDK KGVQLQTHPN VDKKLFTAES LIGLKNPEKS FPVNSDVGVL KWRLQTTEES FIPLTINCWP SESGNGCDVN IEYELQEDNL ELNDVVITIP LPSGVGAPVI GEIDGEYRHD SRRNTLEWCL PVIDAKNKSG SLEFSIAGQP NDFFPVQVSF VSKKNYCNIQ VTKVTQVDGN SPVRFSTETT FLVDKYEIL (164-511 aa encoded by BC093636)
This gene maps in a region, which include the mixed lineage leukemia and Friend leukemia virus integration 1 genes, where multiple disease-associated chromosome translocations occur. It is an intracellular protein. Archain sequences are well conserved among eukaryotes and this protein may play a fundamental role in eukaryotic cell biology. It has similarities to heat shock proteins and clathrin-associated proteins, and may be involved in vesicle structure or trafficking.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Archain; archain vesicle transport protein 1; coatomer delta subunit; coatomer delta-COP; coatomer protein complex, subunit delta; coatomer protein delta-COP; Coatomer subunit delta; Delta-coat protein; Delta-COP; pale coat neuro
Gene Aliases: 4632432M07Rik; ARCN1; COPD; nur17
UniProt ID: (Human) P48444, (Mouse) Q5XJY5
Entrez Gene ID: (Human) 372, (Mouse) 213827
Molecular Function: vesicle coat protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support