Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNIRNAQPD DLMNMQHCNL LCLPENYQMK YYLYHGLSWP QLSYIAEDED GKIVGYVLAK MEEEPDDVPH GHITSLAVKR SHRRLGLAQK LMDQASRAMI ENFNAKYVSL HVRKSNRPAL HLYSNTLNFQ ISEVEPKYYA DGEDAYAMKR DLSQMADELR RQMDLKKGGY VVLGSRENQE TQGSTLSDSE EACQQKNPAT EESGSDSKEP KESVESTNVQ DSSESSDSTS (47-276 aa encoded by BC004552)
Displays alpha (N-terminal) acetyltransferase activity. Proposed alternative catalytic subunit of the N-terminal acetyltransferase A (NatA) complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: =N-terminal acetyltransferase complex ARD1 subunit homolog B; alpha-N-acetyltransferase 1B; ARD1 homolog B; Ard2; arrest defective 2; hARD2; human arrest defective 2; N-alpha-acetyltransferase 11; N-alpha-acetyltransferase 11, NatA catalytic subunit; N-terminal acetyltransferase complex ARD1 subunit homolog B; Nalpha acteyltransferase 11; NatA catalytic subunit Naa11
Gene Aliases: ARD1B; ARD2; C80008; hARD2; NAA11
UniProt ID: (Human) Q6P479, (Mouse) Q3UX61, (Rat) Q4V8K3
Entrez Gene ID: (Human) 84779, (Mouse) 97243, (Rat) 289482
Molecular Function: acetyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support