Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • ARF Pan Antibodies

            Invitrogen

            ARF1/ARF3/ARF5/ARF6 Monoclonal Antibody (1D9)

            Advanced Verification
            1 Published Figure
            18 References
            View all (3) ARF Pan antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # MA3-060
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            516.00
            100 µL
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite ARF1/ARF3/ARF5/ARF6 Monoclonal Antibody (1D9)

            • Antibody Testing Data (13)
            • Published Figures (1)
            • Advanced Verification (1)
            Application
            ARF1/ARF3/ARF5/ARF6 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 15

            ARF1/ARF3/ARF5/ARF6 Antibody (MA3-060) in ICC/IF

            Immunofluorescent analysis of ADP-Ribosylation Factor using ADP-Ribosylation Factor Monoclonal Antibody (1D9) (Product # MA3-060) shows staining in Hela Cells. ADP-Ribosylation Factor (green), F-Actin staining with Phalloidin (red) and nuclei with DAPI (blue) is shown. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with an antibody re... View More
            View Product

            ARF1/ARF3/ARF5/ARF6 Antibody in Immunocytochemistry (ICC/IF)
            ARF1/ARF3/ARF5/ARF6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ARF1/ARF3/ARF5/ARF6 Antibody in Western Blot (WB)
            ARF1/ARF3/ARF5/ARF6 Antibody in Flow Cytometry (Flow)
            ARF1/ARF3/ARF5/ARF6 Antibody in Immunocytochemistry (ICC/IF)
            ARF1/ARF3/ARF5/ARF6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ARF1/ARF3/ARF5/ARF6 Antibody in Immunocytochemistry (ICC/IF)
            ARF1/ARF3/ARF5/ARF6 Antibody in Immunocytochemistry (ICC/IF)
            ARF1/ARF3/ARF5/ARF6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ARF1/ARF3/ARF5/ARF6 Antibody in Western Blot (WB)
            ARF1/ARF3/ARF5/ARF6 Antibody in Western Blot (WB)
            ARF1/ARF3/ARF5/ARF6 Antibody in Flow Cytometry (Flow)
            ARF1/ARF3/ARF5/ARF6 Antibody in Flow Cytometry (Flow)
            ARF1/ARF3/ARF5/ARF6 Antibody in Western Blot (WB)
            ARF1/ARF3/ARF5/ARF6 Antibody

            Product Details

            MA3-060

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:3000
            View 11 publications 11 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:100
            -

            Immunocytochemistry (ICC/IF)

            1:100
            View 5 publications 5 publications

            Flow Cytometry (Flow)

            1/20
            -

            ELISA (ELISA)

            -
            View 1 publication 1 publication

            Immunoprecipitation (IP)

            Assay-dependent
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Dog, Hamster, Human, Mouse, Rat

            Published species

            Bovine, Hamster, Human, Mouse, Non-human primate, Rat

            Host/Isotype

            Mouse / IgG1

            Class

            Monoclonal

            Type

            Antibody

            Clone

            1D9

            Immunogen

            Recombinant human ARF1.

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            Conc. Not Determined

            Storage buffer

            ascites

            Contains

            0.05% sodium azide

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2274419

            Product Specific Information

            MA3-060 detects ADP-ribosylation factor1 (Arf1), Arf3, Arf5 and Arf6 and about ten-fold less well with Arf4 from human, mouse and rat tissues.

            MA3-060 has been successfully used in Western blot, immunofluorescence, immunocytochemistry, and immunoprecipitation procedures. By Western blot, this antibody detects a single ~21 kDa protein representing Arf in rat pancreas extract. Immunofluorescence staining of Arf in rat pancreas with MA3-060 results in staining of the cytoplasmic face of the trans-Golgi membranes. MA3-060 cannot be used to neutralize Arf in solution.

            The MA3-060 antigen is recombinant human Arf1.

            Target Information

            ADP-ribosylation factor 1 (ARF1) is a member of the human ARF gene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6 ARF proteins and 11 ARF-like proteins, constitute a family of the RAS superfamily. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6), and members of each class share a common gene organization. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq, Jul 2008]

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Related Products for

            Western Blot
            • Secondary Antibodies
            • Isotype Controls
            • Loading Controls WB
            • Electrophoresis WB
            • Transfer Stacks WB
            • Substrates WB
            • Labeling Kits WB

            Bioinformatics

            Protein Aliases: ADP-ribosylation factor 1; ADP-ribosylation factor 2; ADP-ribosylation factor 3; ADP-ribosylation factor 4; ADP-ribosylation factor 4-like protein; ADP-ribosylation factor 5; ADP-ribosylation factor 6; ARF; Liver regeneration-related protein LRRG202; pan ARF; small GTP binding protein

            View more View less

            Gene Aliases: 5430400P17Rik; AA407803; Ac1-253; AI788669; AI854770; ARF1; ARF2; ARF3; ARF4; ARF5; ARF6; AW496366; I79_003112

            View more View less

            UniProt ID: (Human) P84077, (Human) P61204, (Human) P18085, (Human) P84085, (Human) P62330, (Mouse) P84078, (Rat) P84079, (Rat) P84082, (Mouse) Q8BSL7, (Mouse) P61205, (Rat) P61206, (Rat) P61751, (Mouse) P61750, (Mouse) P84084, (Rat) P84083, (Mouse) P62331, (Rat) P62332

            View more View less

            Entrez Gene ID: (Dog) 100856517, (Human) 375, (Dog) 486562, (Human) 377, (Dog) 607073, (Human) 378, (Dog) 475206, (Human) 381, (Human) 382, (Dog) 490679, (Chinese hamster) 100757864, (Mouse) 11840, (Rat) 64310, (Rat) 79119, (Mouse) 11841, (Mouse) 11842, (Chinese hamster) 100766631, (Rat) 140940, (Chinese hamster) 100769922, (Rat) 79120, (Mouse) 11843, (Mouse) 11844, (Chinese hamster) 100759676, (Rat) 79117, (Mouse) 11845, (Chinese hamster) 100774654, (Rat) 79121

            View more View less

            Function(s)

            magnesium ion binding GTPase activity receptor signaling protein activity protein binding GTP binding GDP binding poly(A) RNA binding phospholipase D activator activity ARF guanyl-nucleotide exchange factor activity epidermal growth factor receptor binding thioesterase binding protein N-terminus binding nucleotide binding protein homodimerization activity G-protein

            Process(es)

            regulation of receptor internalization phosphatidylinositol biosynthetic process cellular copper ion homeostasis post-Golgi vesicle-mediated transport actin filament organization small GTPase mediated signal transduction protein transport antigen processing and presentation of exogenous peptide antigen via MHC class II regulation of Arp2/3 complex-mediated actin nucleation very-low-density lipoprotein particle assembly positive regulation of catalytic activity positive regulation of endocytosis positive regulation of calcium ion-dependent exocytosis regulation of defense response to virus by virus positive regulation of protein secretion Golgi to transport vesicle transport long term synaptic depression positive regulation of dendritic spine development synaptic vesicle budding dendritic spine organization lysosomal membrane organization positive regulation of sodium ion transmembrane transport positive regulation of late endosome to lysosome transport positive regulation of ER to Golgi vesicle-mediated transport regulation of phospholipid metabolic process retrograde vesicle-mediated transport, Golgi to ER protein ADP-ribosylation ER to Golgi vesicle-mediated transport epidermal growth factor receptor signaling pathway brain development learning cell migration activation of phospholipase D activity negative regulation of apoptotic process positive regulation of GTPase activity apical protein localization establishment or maintenance of epithelial cell apical/basal polarity positive regulation of transcription from RNA polymerase II promoter response to axon injury dendritic spine development protein localization to cilium regulation of reactive oxygen species metabolic process liver development movement of cell or subcellular component cell cycle cell adhesion vesicle-mediated transport positive regulation of actin filament polymerization cortical actin cytoskeleton organization ruffle organization myeloid cell apoptotic process protein localization to cell surface regulation of Rac protein signal transduction protein localization to endosome negative regulation of receptor-mediated endocytosis cell division regulation of filopodium assembly regulation of dendritic spine development positive regulation of establishment of protein localization to plasma membrane establishment of epithelial cell polarity hepatocyte apoptotic process negative regulation of dendrite development transport nervous system development cell differentiation regulation of toll-like receptor 4 signaling pathway actin cytoskeleton organization
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-kfh2r:80/100.66.79.247:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0