Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: SH-SY5Y, U-251MG.
Immunogen sequence: EKKGVYQTLS WKRYQPCWVL MVSVLLHHWK ALKKVNMKLL VNIREREDNV
ARHGAP11B is a Hominin-specific protein that promotes development and evolutionary expansion of the brain neocortex. Able to promote amplification of basal progenitors in the subventricular zone, producing more neurons during fetal corticogenesis. Does not possess GTPase activator activity.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 7, member B1; GAP (1-8); Inactive Rho GTPase-activating protein 11B; Rho-type GTPase-activating protein 11B
Gene Aliases: ARHGAP11B; B'-T; FAM7B1
UniProt ID: (Human) Q3KRB8
Entrez Gene ID: (Human) 89839
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support