Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VCDVVLNHA PDFRRVYLPY VTNQTYQERT FQSLMNSNSN FREVLEKLES DPVCQRLSLK SFLILPFQRI TRLKLLLQNI LKRTQPGSSE EAEATKAHHA LEQLIRDCNN NVQSMRRTEE LIYLSQKIEF ECKIFPLISQ SRWLVKSGEL TALEFSASPG LRRKLNTRPV HLHLFNDCLL LSRPREGSRF LVFDHAPFSS IRGEKCEMKL HGPHKNLFRL FLRQNTQGAQ AEFLFRTETQ SEKLRWISAL AMPREELDLL ECYNSPQVQC LRAYKPREND ELALEKADVV MVTQQSSDGW LEGVRLSDGE RGWFPVQQVE FISNPEVRAQ NLKEAHRVKT AKLQLVEQQA (171-519 aa encoded by BC010046)
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARH5; DKFZp686N1969; Ephexin 3; Ephexin-3; Guanine nucleotide regulatory protein TIM; Oncogene TIM; p60 TIM; Rho guanine nucleotide exchange factor (GEF) 5; Rho guanine nucleotide exchange factor 5; Transforming immortalized mammary oncogene
Gene Aliases: 2210412D05Rik; AA717842; ARHGEF5; AW495314; GEF5; P60; TIM; TIM1
UniProt ID: (Human) Q12774, (Mouse) E9Q7D5
Entrez Gene ID: (Human) 7984, (Mouse) 54324, (Rat) 140898
Molecular Function:
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support