Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KELELQILT EAIRNWEGDD IKTLGNVTYM SQVLIQCAGS EEKNERYLLL FPNVLLMLSA SPRMSGFIYQ GKLPTTGMTI TKLEDSENHR NAFEISGSMI ERILVSCNNQ QDLQEWVEHL QKQTKVTSVG NPTIKPHSVP SHTLPSHPVT PSSKHADSKP APLTPAYHTL PHPSHHGTPH TTINWGPLEP PKTPKPWSLS CLRPAPPLRP SAALCYKEDL SKSPKTMKKL LPKRKPERKP SDEEFASRKS TAALEEDAQI LKVIEAYCTS AKTRQTLNST WQGTDLMHNH VLADDDQPSL DSLGRRSSLS RLEPSDLSED SDYDSIWTAH SYRMGSTSRK SCCSYISHQN (455-803 aa encoded by BC060776)
The serine/threonine kinase, p21 activated kinase (PAK), is a downstream effector of the small GTPases Cdc42 and Rac. PAK associates with Nck, the p85 and p110 subunits of PI3-kinase and PIX (PAK-interacting exchange factor) in a focal complex. The binding of PIX is necessary for the localization and activation of PAK in the Cdc42 to Rac signaling pathway, and this binding occurs through the high affinity of the N-terminal SH3 domain of PIX for a conserved proline rich PAK sequence. PIX exists as two isoforms, alpha and beta, and both are highly expressed in heart, muscle and thymus tissues of human and rat. alphaPIX is phosphorylated via PDGF and EphB2 receptor signaling pathways or through association with PI3-kinase. The alphaPIX isoform predominantly acts as a guanine nucleotide exchange factor (GEF) on Rac, which may mediate lamellipodia formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-Pix; COOL-1; p85; p85SPR; PAK-interacting exchange factor beta; Pak3 binding protein; PIXbeta; Rho guanine nucleotide exchange factor (GEF) 7; Rho guanine nucleotide exchange factor 7; SH3 domain-containing proline-rich protein
Gene Aliases: ARHGEF7; BETA-PIX; betaPix; betaPix-b; betaPix-c; Cool; COOL-1; COOL1; KIAA0142; mKIAA0142; Nbla10314; P50; P50BP; P85; P85COOL1; P85SPR; PAK3; PAK3BP; PIX; PIXB
UniProt ID: (Human) Q14155, (Rat) O55043, (Mouse) Q9ES28
Entrez Gene ID: (Human) 8874, (Rat) 114559, (Mouse) 54126
Molecular Function:
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support