Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Sequence of this protein is as follows: MTDNSNNQLV VRAKFNFQQT NEDELSFSKG DVIHVTRVEE GGWWEGTLNG RTGWFPSNYV REVKASEKPV SPKSGTLKSP PKGFDTTAIN KSYYNVVLQN ILETENEYSK ELQTVLSTYL RPLQTSEKLS SANISYLMGN LEEICSFQQM LVQSLEECTK LPEAQQRVGG CFLNLMPQMK TLYLTYCANH PSAVNVLTEH SEELGEFMET KGASSPGILV LTTGLSKPFM RLDKYPTLLK ELERHMEDYH TDRQDIQKSM AAFKNLSAQC QEVRKRKELE LQILTEAIRN WEGDDIKTLG NVTYMSQVLI QCAGSEEKNE RYLLLFPNVL LMLSASPRMS GFIYQGKLPT TGMTITKLED SENHRNAFEI SGSMIERILV SCNNQQDLQE WVEHLQKQTK VTSVGNPTIK PHSVPSHTLP SHPVTPSSKH ADSKPAPLTP AYHTLPHPSH HGTPHTTINW GPLEPPKTPK PWSLSCLRPA PPLRPSAALC YKEDLSKSPK TMKKLLPKRK PERKPSDEEF ASRKSTAALE EDAQILKVIE AYCTSAKTRQ TLNSSSRKES APQVLLPEEE KIIVEETKSN GQTVIEEKSL VDTVYALKDE VQELRQDNKK MKKSLEEEQR ARKDLEKLVR KVLKNMNDPA WDETNL
The serine/threonine kinase, p21 activated kinase (PAK), is a downstream effector of the small GTPases Cdc42 and Rac. PAK associates with Nck, the p85 and p110 subunits of PI3-kinase and PIX (PAK-interacting exchange factor) in a focal complex. The binding of PIX is necessary for the localization and activation of PAK in the Cdc42 to Rac signaling pathway, and this binding occurs through the high affinity of the N-terminal SH3 domain of PIX for a conserved proline rich PAK sequence. PIX exists as two isoforms, alpha and beta, and both are highly expressed in heart, muscle and thymus tissues of human and rat. alphaPIX is phosphorylated via PDGF and EphB2 receptor signaling pathways or through association with PI3-kinase. The alphaPIX isoform predominantly acts as a guanine nucleotide exchange factor (GEF) on Rac, which may mediate lamellipodia formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-Pix; COOL-1; p85; PAK-interacting exchange factor beta; PIXbeta; Rho guanine nucleotide exchange factor (GEF) 7; Rho guanine nucleotide exchange factor 7; SH3 domain-containing proline-rich protein
Gene Aliases: ARHGEF7; BETA-PIX; COOL-1; COOL1; KIAA0142; Nbla10314; P50; P50BP; P85; P85COOL1; P85SPR; PAK3; PAK3BP; PIXB
UniProt ID: (Human) Q14155
Entrez Gene ID: (Human) 8874
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support