Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 14
Keep as concentrated solution.
Predicted reactivity: Mouse (95%).
Positive Control: 293T, A431, HeLa, HepG2, HeLa nuclear, human ARID2-transfected 293T.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ARID2 is a subunit of the PBAF chromatin-remodeling complex which facilitates ligand-dependent transcriptional activation by nuclear receptors.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arid 2; ARID domain-containing protein 2; AT rich interactive domain 2 (ARID, RFX-like); AT-rich interactive domain-containing protein 2; BAF200; BRG1-associated factor 200; RFX-like; Zinc finger protein with activation potential; zipzap; Zipzap/p200
Gene Aliases: ARID2; BAF200; KIAA1557; p200
UniProt ID: (Human) Q9HCL5
Entrez Gene ID: (Human) 196528
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support