Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: FLDDLFSFMQ KRGTPVNRIP IMAKQVLDLF MLYVLVTEKG GLVEVINKKL WREITKGLNL PTSITSAAFT LRTQYMEYLY PYECEKRGLS NPNELQAAID SNRREGRRQS FGGSLFAYSP GGAHGMLSSP KLPVSSLGLA ASTNGSSITP APKIKKEEDS AIPITVPGRL PVSLAGHPVV AAQAAAVQAA AAQAAVAAQA AALEQLREKL ESAEPPEKKM ALVADEQQRL MQRALQQNFL AMAAQLPMSI RINSQASESR QDSAVNLTGT NGSNSISMSV EINGIMYTGV LFAQPPAPTP TSAPNKGGGG GGSSSSNAGG RGGNTGTSGG QAGPAGLSTP STSTSNNSLP
The mouse Bright (B-cell regulator of IgH transcription) protein is expressed specifically in B cells and transactivates an IgH enhancer in transient transfection assays. Bright and the Drosophila 'dead ringer' (dri) gene product belong to a protein family whose members share a conserved DNA-binding domain termed the 'A/T-rich interaction domain' (ARID).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARID domain-containing 3A; ARID domain-containing protein 3A; AT rich interactive domain 3A (BRIGHT- like) protein; AT rich interactive domain 3A (BRIGHT-like); AT-rich interactive domain-containing protein 3A; B-cell regulator of IgH transcription; Bright; dead ringer-like 1; Dead ringer-like protein 1; E2F-binding protein 1
Gene Aliases: ARID3A; BRIGHT; DRIL1; DRIL3; DRX; E2FBP1
UniProt ID: (Human) Q99856
Entrez Gene ID: (Human) 1820
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support