Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AEESQEGLCE RESANGFETN VASGTCSIIV QERESREKGQ KRPSDGNSGL MAKKQKRTPK RTSAAAKNEK NGTGQSSDSE DLPVLDNSSK CTPVKHLNVS KPQKLAR
ARID4A encodes a ubiquitously expressed nuclear protein. It binds directly, with several other proteins, to retinoblastoma protein (pRB) which regulates cell proliferation. pRB represses transcription by recruiting the encoded protein. This protein, in turn, serves as a bridging molecule to recruit HDACs and, in addition, provides a second HDAC-independent repression function. The encoded protein possesses transcriptional repression activity. Multiple alternatively spliced transcripts have been observed for this gene, although not all transcript variants have been fully described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARID domain-containing protein 4A; AT rich interactive domain 4A (RBP1-like); AT-rich interactive domain-containing protein 4A; RBBP-1; retinoblastoma binding protein 1; Retinoblastoma-binding protein 1
Gene Aliases: ARID4A; RBBP-1; RBBP1; RBP-1; RBP1
UniProt ID: (Human) P29374
Entrez Gene ID: (Human) 5926
Molecular Function: transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support