Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The exact function of C18orf25 remains unknown. There are two named isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 180 kDa Sin3-associated polypeptide; ARID domain-containing protein 4B; AT rich interactive domain 4B (RBP1-like); AT-rich interactive domain-containing protein 4B; breast cancer-associated antigen 1; Breast cancer-associated antigen BRCAA1; breast carcinoma-associated antigen; Histone deacetylase complex subunit SAP180; Rb-binding protein homolog; Retinoblastoma-binding protein 1-like 1; Sin3-associated polypeptide p180; SIN3A-associated protein 180
Gene Aliases: ARID4B; BCAA; BRCAA1; RBBP1L1; RBP1L1; SAP180
UniProt ID: (Human) Q4LE39
Entrez Gene ID: (Human) 51742
Molecular Function: transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support