Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, mouse brain.
Predicted reactivity: Mouse (99%), Rat (100%), Zebrafish (93%), Xenopus laevis (98%), Chicken (99%), Chimpanzee (99%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
E3 ubiquitin-protein ligase mediating 'Lys-48'-and 'Lys-63'-linked polyubiquitination and subsequent proteasomal degradation of modified proteins. May play a role in myelopoiesis.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: all-trans retinoic acid inducible RING finger; ARI-2; ariadne 2; ariadne homolog 2; E3 ubiquitin-protein ligase ARIH2; FLJ10938; FLJ33921; OTTHUMP00000210388; protein ariadne-2 homolog; RING-type E3 ubiquitin transferase ARIH2; Triad1 protein; UbcM4-interacting protein 48
Gene Aliases: AI843547; ARI2; ARIH2; HT005; TRIAD1; UIP48
UniProt ID: (Human) O95376, (Mouse) Q9Z1K6
Entrez Gene ID: (Human) 10425, (Mouse) 23807
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support