Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
E3 ubiquitin-protein ligase mediating 'Lys-48'-and 'Lys-63'-linked polyubiquitination and subsequent proteasomal degradation of modified proteins. May play a role in myelopoiesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: all-trans retinoic acid inducible RING finger; ARI-2; ariadne homolog 2; E3 ubiquitin-protein ligase ARIH2; FLJ10938; FLJ33921; OTTHUMP00000210388; protein ariadne-2 homolog; Triad1 protein
Gene Aliases: ARI2; ARIH2; HT005; TRIAD1
UniProt ID: (Human) O95376
Entrez Gene ID: (Human) 10425
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support