Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ARL8 belongs to a family of proteins that are structurally similar to ADP-ribosylation factors. ARLs and ARFs are part of the RAS superfamily of regulatory GTPases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor like GTPase 5B; ADP-ribosylation factor-like 5B; ADP-ribosylation factor-like 8; ADP-ribosylation factor-like protein 5B; ADP-ribosylation factor-like protein 8; ADP-ribosylation-like factor 8
Gene Aliases: ARL5B; ARL8
UniProt ID: (Human) Q96KC2
Entrez Gene ID: (Human) 221079
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support