Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Immunogen sequence: MILLEVNNRI IEETLALKFE NAAAGNKPEA VEVTFADFDG VLYHISNPNG DKTKVMVSIS LKFYKELQAH GADELLKRVY GSFLVNPESG YNVSLLYDLE NLPASKDSIV HQAGMLKRNC FASVFEKYFQ FQEEGKEGEN RAVIHYRDDE TMYVESKKDR VTVVFSTVFK DDDDVVIGKV FMQEFKEGRR ASHTAPQVLF SHREPPLELK DTDAAVGDNI GYITFVLFPR HTNASARDNT INLIHTFRDY LHYHIKCSKA YIHTRMRAKT SDFLKVLNRA RPDAEKKEMK TITGKTFSSR
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p34 subunit, has yet to be determined. Two alternatively spliced variants have been characterized to date. Additional alternatively spliced variants have been described but their full length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin related protein 2/3 complex subunit 2, 34kDa; actin related protein 2/3 complex, subunit 2, 34kDa; Actin-related protein 2/3 complex subunit 2; actin-related protein 2/3 complex subunit 2 {ECO:0000250|UniProtKB:O15144}; Arp2/3 complex 34 kDa subunit; arp2/3 complex 34 kDa subunit {ECO:0000250|UniProtKB:O15144}; ARP2/3 protein complex subunit 34; arpc2 {ECO:0000250|UniProtKB:O15144}; p34-ARC; p34-ARC {ECO:0000250|UniProtKB:O15144}; testis tissue sperm-binding protein Li 53e
Gene Aliases: 2210023N03Rik; 34kDa; ARC34; ARPC2; p34-Arc; PNAS-139; PRO2446
UniProt ID: (Human) O15144, (Mouse) Q9CVB6
Entrez Gene ID: (Human) 10109, (Rat) 301511, (Mouse) 76709
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support